missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3RF1 Monoclonal Antibody (K1E030_5E8), Invitrogen™
Mouse Monoclonal Antibody
€ 493.00 - € 868.00
Specifications
| Antigen | SH3RF1 |
|---|---|
| Clone | K1E030_5E8 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30199343
|
Invitrogen™
MA555676 |
50 μL |
€ 493.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30196193
|
Invitrogen™
MA556056 |
200 μL |
€ 868.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: GNSSATKPDK DSKKEKKGLL KLLSGASTKR KPRVSPPASP TLEVELGSAE LPLQGAVGPE LPPGGGHGRA GSCPVDGDGP VTTAVAGAAL AQDAFHRKAS SLDSAVPIAP PPRQACSSLG PVLNESRPVV CERHRVVVSY PPQSEAELEL KEGDIVFVHK KREDGWFKGT LQRNGKTGLF
SH3RF1 Monoclonal antibody specifically detects SH3RF1 in Human, Mouse samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| SH3RF1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2200003J05Rik; E3 ubiquitin-protein ligase SH3RF1; FLJ21602; KIAA1494; plenty of SH3 domains; plenty of SH3s; Posh; Posh1; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative E3 ubiquitin-protein ligase SH3RF1; E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; R75531; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; RNF142; SH3 domain containing ring finger 1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2; SH3MD2; Sh3rf1 | |
| Recombinant human SH3RF1 (immunogen range is 701-880). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SH3RF1 |
| K1E030_5E8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q69ZI1, Q7Z6J0 | |
| 57630, 59009 | |
| IgG2a | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title