missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH3RF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16301915
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16301915 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16301915 Supplier Invitrogen™ Supplier No. PA584961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AVTNASQAKV PMSTAGQTSR GVTMVSPSTA GGPAQKLQGN GVAGSPSVVP AAVVSAAHIQ TSPQAKVLLH Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 94%.

This gene encodes a protein containing an N-terminus RING-finger, four SH3 domains, and a region implicated in binding of the Rho GTPase Rac. Via the RING-finger, the encoded protein has been shown to function as an ubiquitin-protein ligase involved in protein sorting at the trans-Golgi network. The encoded protein may also act as a scaffold for the c-Jun N-terminal kinase signaling pathway, facilitating the formation of a functional signaling module.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH3RF1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH3RF1
Gene Accession No. Q7Z6J0
Gene Alias 2200003J05Rik; E3 ubiquitin-protein ligase SH3RF1; FLJ21602; KIAA1494; plenty of SH3 domains; plenty of SH3s; Posh; Posh1; Protein POSH; putative E3 ubiquitin-protein ligase SH3RF1; putative E3 ubiquitin-protein ligase SH3RF1; E3 ubiquitin-protein ligase SH3RF1; putative scaffolding protein POSH; R75531; RING finger protein 142; RING-type E3 ubiquitin transferase SH3RF1; RNF142; SH3 domain containing ring finger 1; SH3 domain-containing RING finger protein 1; SH3 multiple domains 2; SH3 multiple domains protein 2; SH3MD2; Sh3rf1
Gene Symbols SH3RF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH3RF1. Recombinant protein control fragment (Product #RP-108017).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57630
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.