Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II, White
Shop All Epredia Products
Click to view available options
Color:
Lilac
White
Unit Size:
Each
Pack of 500
Description
- Acetal polymer construction
- Locking lid attached by two hinges
- Cassette writing face is at a 45° angle
- Pore style is small square
- Variety of colors available
Ordering Alerts:
- Supplied in boxes of 500 cassettes with attached lids
Specifications
Specifications
| Pore Style | Small Square Pore |
| Writing Face Angle | 45° |
| Color | White |
| Material | Acetal Polymer |
| Product Type | Biopsy Cassette |