Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II, White
Shop All Epredia Products
Click to view available options
Color:
Lilac
White
Förpackningsstorlek
Each
Pack of 500
Beskrivelse
- Acetal polymer construction
- Locking lid attached by two hinges
- Cassette writing face is at a 45° angle
- Pore style is small square
- Variety of colors available
Ordering Alerts:
- Supplied in boxes of 500 cassettes with attached lids
Tekniske data
Tekniske data
| Pore Style | Small Square Pore |
| Writing Face Angle | 45° |
| Color | White |
| Material | Acetal Polymer |
| Product Type | Biopsy Cassette |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion