Learn More
Epredia™ Cryomatrix™ embedding resin
Product Code. 12868290 Shop All Epredia Products
Change view
Click to view available options
Capacity:
120 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
12868290 120 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12868290 Supplier Epredia™ Supplier No. 6769006
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Secure and support tissue with Epredia™ Cryomatrix™ embedding resin, an outstanding product used to support tissue for frozen sectioning.
  • Forms a firm bond with the specimen holder
  • Freezes quickly, sections smoothly
  • Dissolves in water without residue
TRUSTED_SUSTAINABILITY
Specifications
Capacity 120 mL
Product Type Embedding Media