Learn More
Epredia™ Shandon™ EA-65 Stain
Product Code. 12647936 Shop All Epredia Products
Click to view available options
Quantity:
4L
2 x 1L
Unit Size:
Each
Pack of 2
Product Code. 12647936
Brand: Epredia™ 6766004
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.
  • Polychromatic cytoplasmic stain
  • Ideal for thicker non-GYN preparations
  • Biological Stain Commission-certified dye
TRUSTED_SUSTAINABILITY
Specifications
Type Cellular Components Stain
Quantity 4L