Description
- Ready-to-use
- Produces consistent results
- Stable and precipitate-free; no filtering
- Compatible with all alum hematoxylin stains
- Available in alcoholic or aqueous formulations
Specifications
Specifications
| Type | Alcoholic Eosin Y Stain |
| Quantity | 1L |