Learn More
Epredia™ Shandon™ Para-Release™ Spray
Ease the release of paraffin blocks from embedding molds with Epredia™ Shandon™ Para-Release™ Spray.
Brand: Epredia™ 9990420
This item is not returnable.
View return policy
Description
- Apply spray-releasing agent to molds before adding paraffin for clean release of block.
Specifications
50 mL |