missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-167R
This item is not returnable.
View return policy
Description
SHANK1 Polyclonal antibody specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SHANK1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
| residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. | |
| 0.1 mL | |
| Neuroscience | |
| 50944 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction