Learn More
Invitrogen™ SHANK1 Monoclonal Antibody (N22/21), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545447
Description
1 μg/mL of MA5-45447 was sufficient for detection of Shank1 in 10 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 190-220 kDa (alternative splice variants). No cross-reactivity against Shank2 or Shank3.
Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. May play a role in the structural and functional organization of the dendritic spine and synaptic junction.
Specifications
| SHANK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
| D3YZU1, Q9WV48, Q9Y566 | |
| SHANK1 | |
| Fusion protein amino acids 469-691 (SH3/PDZ domains) of rat Shank1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray | |
| N22/21 | |
| FITC | |
| SHANK1 | |
| GKAP/SAPAP interacting protein; GKAP/SAPAP-interacting protein; OTTHUMP00000174437; SAPAP-interacting protein synamon; SH3 and multiple ankyrin repeat domains 1; SH3 and multiple ankyrin repeat domains protein 1; SH3/ankyrin domain gene 1; SHANK 1; Shank1; Shank1a; somatostatin receptor-interacting protein; SPANK 1; Spank1; SPANK-1; SSTR-interacting protein; Sstrip; Synamon | |
| Mouse | |
| Protein G | |
| RUO | |
| 243961, 50944, 78957 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.