missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41171-0.025mg
This item is not returnable.
View return policy
Description
SHANK3 Polyclonal specifically detects SHANK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SHANK3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SHANK3 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human SHANK3. The immunogen is located within amino acids 890 - 940 of SHANK3. Amino Acid Squence: APRPPPAATPPERPKRRPR | |
| 0.025 mg | |
| Primary | |
| At least three alternatively spliced transcript isoforms of SHANK3 are known to exist. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 85358 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction