missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHANK3 Antibody (S69), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47610JF646
This item is not returnable.
View return policy
Description
SHANK3 Monoclonal antibody specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Microarray
Specifications
| SHANK3 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2 | |
| Synthetic peptide amino acids 840-857 of rat Shank3 | |
| 0.1 mL | |
| Neuroscience | |
| 85358 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Microarray | |
| S69 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction