Learn More
B Braun™ Medibox™ Sharps Disposal Container
Click to view available options
Capacity (Metric):
0.6 L
Unit Size:
Pack of 10
Description
Medibox sharps containers are compliant with international standards
- Impact and puncture resistant
- Freestanding, ergonomic design for easy handling
- Handling instructions by self-explaining pictograms
- Overfill warning by maximum fill line and visible content level
- Diverse ports for disposal without contact
Specifications
Specifications
| Capacity (Metric) | 0.6 L |
| Certifications/Compliance | ISO 23907, ISO 9001 |
| Depth (Metric) | 75 mm |
| Color | White with Yellow Label |
| Height (Metric) | 130 mm |
| Quantity | 1 Ea. |
| Width (Metric) | 116 mm |
| Material | Polypropylene |