missing translation for 'onlineSavingsMsg'
Learn More

SHB Antibody (SR1974), Novus Biologicals™

Product Code. p-200006132 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18670785 100 μg 100µL
18602575 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18670785 Supplier Novus Biologicals Supplier No. NBP322624100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SHB Monoclonal antibody specifically detects SHB in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHB
Applications Western Blot
Classification Monoclonal
Clone SR1974
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias bA3J10.2, RP11-3J10.8, SH2 domain-containing adapter protein B, SHB (Src homology 2 domain containing) adaptor protein B, SHB adaptor protein (a Src homology 2 protein), Src homology 2 domain containing adaptor protein B
Host Species Rabbit
Immunogen A synthesized peptide derived from human SHB (Uniprot #: Q15464)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6461
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.