missing translation for 'onlineSavingsMsg'
Learn More

SHBG Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 178.00 - € 507.00

Specifications

Antigen SHBG
Concentration 1 mg/mL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18637676
View Documents
Novus Biologicals
NBP2-41294-0.025mg
0.025 mg
€ 178.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18716753
View Documents
Novus Biologicals
NBP2-41294
0.1 mg
€ 507.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SHBG Polyclonal specifically detects SHBG in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

SHBG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse, Rat
ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
SHBG
IgG
Affinity Purified
35 kDa
1 mg/mL
Polyclonal
Rabbit
Cancer, Vision
PBS with 0.02% Sodium Azide
6462
Antibody was raised against a 16 amino acid peptide near the center of human SHBG. The immunogen is located within amino acids 220 - 270 of SHBG. Amino Acid Squence: AEFNLRDIPQPHAEPW
Primary
At least three isoforms of SHBG are known to exist; this antibody will detect all three isoforms.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.