missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHBG Antibody (OTI1H10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74166V
This item is not returnable.
View return policy
Description
SHBG Monoclonal specifically detects SHBG in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SHBG | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SHBG | |
| Full length recombinant protein of human SHBG (NP_001139751) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI1H10 | |
| Western Blot, Immunohistochemistry | |
| ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
| Mouse | |
| Affinity Purified | |
| Cancer, Vision | |
| 6462.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction