missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHBG Antibody (SPM605), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11425PCP
This item is not returnable.
View return policy
Description
SHBG Monoclonal antibody specifically detects SHBG in Human samples. It is validated for Flow Cytometry
Specifications
| SHBG | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| SPM605 | |
| Flow Cytometry | |
| ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin | |
| Recombinant full-length human SHBG protein (Uniprot: P04278) | |
| 0.1 mL | |
| Cancer, Vision | |
| 6462 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction