missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHBG Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507184
This item is not returnable.
View return policy
Description
SHBG is a steroid binding protein that was first described as a plasma protein secreted by the liver and is thought to participate in the regulation of steroid responses. SHBG transports androgens and estrogens in the blood, binding each steroid molecule as a dimer formed from identical or nearly identical monomers. Low plasma SHBG levels are associated with obesity, abdominal adiposity, metabolic syndrome, and predict the development of type 2 diabetes. Polymorphisms in this gene have been associated with polycystic ovary syndrome and type 2 diabetes mellitus.Specifications
| SHBG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04278 | |
| SHBG | |
| Full length human recombit protein of human SHBG produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| SHBG | |
| ABP, SBP, TEBG | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6462 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction