missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHBG Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA507184S
This item is not returnable.
View return policy
Description
SHBG Monoclonal antibody specifically detects SHBG in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| SHBG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04278 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6462 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunocytochemistry, Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| SHBG | |
| ABP; Abpa; androgen binding protein; Androgen binding protein (ABP); SBP; sex hormone binding globulin; Sex hormone binding globulin or androgen-binding protein (other gene product from ABP gene); sex hormone-binding globulin; Sex hormone-binding globulin (SHBG); Sex steroid binding protein (SBP); Sex steroid-binding protein; Shbg; TeBG; testis-specific androgen-binding protein; Testosterone binding beta globulin; testosterone-binding beta-globulin; testosterone-estradiol-binding globulin; Testosterone-Estradiol-Binding Globulin (TEBG); Testosterone-estrogen-binding globulin | |
| Full length human recombinant protein of human SHBG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction