missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC1 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501074S
This item is not returnable.
View return policy
Description
SHC1 Monoclonal antibody specifically detects SHC1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| SHC1 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23469 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5791 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1D3 | |
| Unconjugated | |
| PTPRE | |
| FLJ26504; OTTHUMP00000035472; p66; P66shc; RP11-307C12.1; SH2 domain protein C1; SHC; SHC (Src homology 2 domain containing) transforming protein 1; SHC (Src homology 2 domain-containing) transforming protein 1; SHC adaptor protein 1; SHC1; SHCA; SHC-transforming protein 1; SHC-transforming protein 3; SHC-transforming protein A; src homology 2 domain-containing transforming protein C1; src homology 2 domain-containing-transforming protein C1 | |
| Full length human recombinant protein of human PTPRE produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction