missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13307-25ul
This item is not returnable.
View return policy
Description
SHC3 Polyclonal antibody specifically detects SHC3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SHC3 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Neuronal Shc, NSHCDKFZp686H1544, N-Shcsrc homology 2 domain containing transforming protein C3, Protein Rai, RAI, SH2 domain protein C3, SHC (Src homology 2 domain containing) transforming protein 3, SHCCFLJ45325, SHC-transforming protein 3, SHC-transforming protein C, src homology 2 domain-containing transforming protein C3, Src homology 2 domain-containing-transforming protein C3 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: HPYYNSIPSKMPPPGGFLDTRLKPRPHAPDTAQFAGKEQTYYQGRHLGDTFGEDWQQTPLRQGSSDIYSTPEGKLHVAPTGEAPTYVNTQQIPP | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 53358 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction