missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SHC3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16602705 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16602705 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16602705 Supplier Invitrogen™ Supplier No. PA5106262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Shc3.

Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHC3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SHC3
Gene Accession No. O70143, Q61120, Q92529
Gene Alias neuronal Shc; Nshc; N-Shc; Protein Rai; RAI; SH2 domain protein C3; SHC (Src homology 2 domain containing) transforming protein 3; SHC adaptor protein 3; SHC3; Shc3 p51; SHCC; SHC-transforming protein 3; SHC-transforming protein C; src homology 2 domain-containing transforming protein C3; src homology 2 domain-containing-transforming protein C3
Gene Symbols SHC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SHC3(Accession Q92529), corresponding to amino acid residues L398-K448.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114858, 20418, 53358
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.