missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567331
This item is not returnable.
View return policy
Description
Immunogen sequence: SQESPTPLCTL IPRMASMKLA NPATLLSLKN FCLGTKEVPR LKLQESRDPG SSGPSS Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.
Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.
Specifications
| SHC4 | |
| Polyclonal | |
| Unconjugated | |
| SHC4 | |
| 6230417E10Rik; 9930029B02Rik; Gm685; hShcD; mShcD; novel phosphotyrosine interaction domain (PTB/PID) and SH2 domains containing protein; rai-like protein; rai-like protein RaLP; RaLP; RGD1309182; SH2 domain protein C4; SHC (Src homology 2 domain containing) family member 4; SHC (Src homology 2 domain containing) family, member 4; SHC adaptor protein 4; Shc4; Shcd; SHC-transforming protein 4; SHC-transforming protein D; src homology 2 domain-containing-transforming protein C4; UNQ6438/PRO21364 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 399694 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6S5L8 | |
| SHC4 | |
| Recombinant Human SHC4. Recombinant protein control fragment (Product #RP-107979). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction