Learn More
BCL2, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against BCL2.
Brand: Abnova PAB9708.250ug
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Specifications
| BCL2 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against BCL2. | |
| In PBS (0.08% sodium azide) | |
| Bcl-2 | |
| Sheep | |
| 250 μg | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:5000) Immunofluorescence (1:5000) The optimal working dilution should be determined by the end user. | |
| BCL2 | |
| BCL2 | |
| Human BCL2. | |
| RUO | |
| 596 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only