Learn More
Sheep anti-Bovine IgG (H+L) Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NB775
Product Code. 18030806
  • € 264.00 / 1mL
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Unconjugated
Sheep
Affinity Purified
RUO
Secondary
Bovine
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 1:1000-1:10000, ELISA, Immunohistochemistry 1:200 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:2000, Immunohistochemistry-Paraffin
Bovine IgG-heavy and light chain
1 mL
Adaptive Immunity, Immunology
By immunoelectrophoresis and ELISA this reacts specifically with bovine IgG and with light chains common to other bovine immunoglobulins. No was detected against non-immunoglobulin serum proteins. This may cross react with IgG from other species.
Store at 4C. Do not freeze.