missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG Heavy Chain Secondary Antibody, DyLight 405, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB790V
This item is not returnable.
View return policy
Description
IgG Heavy Chain Polyclonal antibody specifically detects IgG Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Bovine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| Immunoglobulin G, ImmunoglobulinG | |
| Bovine IgG-heavy chain | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction