missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG1 Heavy Chain Secondary Antibody, DyLight 594, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB780DL594
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Polyclonal antibody specifically detects IgG1 Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG1 Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Bovine IgG1 | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction