missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG1 Secondary Antibody, Janelia Fluor 646, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NBP2-68328JF646
This item is not returnable.
View return policy
Description
IgG1 Polyclonal antibody specifically detects IgG1 in Bovine samples. It is validated for ELISA, Immunodiffusion
Specifications
| IgG1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Sheep | |
| Affinity purified | |
| RUO | |
| Bovine | |
| Purified |
| ELISA, Immunodiffusion | |
| Janelia Fluor 646 | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Purified bovine IgG1 | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction