missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Bovine IgG2 Heavy Chain Secondary Antibody, DyLight 680, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB785FR
This item is not returnable.
View return policy
Description
IgG2 Heavy Chain Polyclonal antibody specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2 Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Bovine IgG2 | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 680 | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction