missing translation for 'onlineSavingsMsg'
Learn More

Bub1b Sheep anti-Mouse, Polyclonal Antibody, Abnova™

Product Code. 16021908
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16021908 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16021908 Supplier Abnova Supplier No. PAB9784.250ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against partial recombinant Bub1b.

Specifications

Antigen Bub1b
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against partial recombinant Bub1b.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Bub1b
Gene Alias AU045529/BUBR1/MGC36092
Gene Symbols Bub1b
Host Species Sheep
Immunogen Recombinant protein corresponding to amino acids 10-190 of mouse Bub1b.
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12236
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.