missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Canine IgG2 Heavy Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB7334-0.5mg
This item is not returnable.
View return policy
Description
IgG2 Heavy Chain Polyclonal antibody specifically detects IgG2 Heavy Chain in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgG2 Heavy Chain | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| 50 mM HEPES pH 7.1, 0.1 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2 containing 0.2%BSA | |
| Dog IgG2 | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:1000 - 1:10000, ELISA, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction