missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E1A, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against recombinant E1A.
Brand: Abnova PAB13560.100ug
This item is not returnable.
View return policy
Specifications
| E1A | |
| Polyclonal | |
| Sheep polyclonal antibody raised against recombinant E1A. | |
| In PBS (0.08% sodium azide) | |
| Recombinant GST fusion protein corresponding to Adenovirus E1A. | |
| RUO | |
| Adenovirus | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| Sheep | |
| 100 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction