missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F5 Sheep anti-Bovine, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against native F5.
Brand: Abnova PAB0665.1mg
This item is not returnable.
View return policy
Specifications
| F5 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against native F5. | |
| In PBS (50% glycerol) | |
| F5 | |
| Native purified Bovine F5. | |
| RUO | |
| 280687 | |
| Bovine | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (25 ug/mL) The optimal working dilution should be determined by the end user. | |
| F5 | |
| Sheep | |
| 1 mg | |
| Primary | |
| This antibody recognizes bovine factor V. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction