missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Human Lambda Light Chain Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB7482H
This item is not returnable.
View return policy
Description
Lambda Light Chain Polyclonal antibody specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Lambda Light Chain | |
| Polyclonal | |
| PBS | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| HRP | |
| IGLC, immunoglobulin lambda constant group | |
| Human Lambda Free Light Chain | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| 3537 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction