Learn More
Sheep anti-Mouse IgG (H+L) Secondary Antibody, PE/Cy7, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NB120-6710PECY7
Product Code. 18510691
  • € 822.00 / 500µL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
PBS
Sheep anti-Mouse IgG (H+L) Secondary Antibody was produced by repeated immunization with Mouse IgG whole molecule in sheep.
500 μg
Adaptive Immunity, Immunology
Mouse
IgG
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Dot Blot
PE-Cyanine7
Sheep
Multi-step
RUO
Secondary
Store at 4°C in the dark. Do not freeze.