Learn More
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Janelia Fluor 549, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72695JF549
Product Code. 18438143
  • € 368.00 / 0.50mL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
50mM Sodium Borate
Rabbit IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Rabbit
IgG
Western Blot, ELISA, Immunohistochemistry, Dot Blot
Janelia Fluor 549
Sheep
Multi-step
RUO
Secondary
Store at 4°C in the dark.