missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A1 Sheep anti-Bovine, Pig, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against S100A1.
Brand: Abnova PAB17075.200uL
This item is not returnable.
View return policy
Specifications
| S100A1 | |
| Polyclonal | |
| Sheep polyclonal antibody raised against S100A1. | |
| In PBS | |
| Pig S100A1. | |
| RUO | |
| Crossreactivity with other members of the S100 family has not been investigated. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot | |
| Unconjugated | |
| Western Blot (1:50) The optimal working dilution should be determined by the end user. | |
| Sheep | |
| 200 μL | |
| Primary | |
| Bovine, Pig | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction