missing translation for 'onlineSavingsMsg'
Learn More

Thromboxane B2, Sheep, Polyclonal Antibody, Abnova™

Product Code. 16080847
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16080847 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16080847 Supplier Abnova Supplier No. PAB13667.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep polyclonal antibody raised against Thromboxane B2.

Specifications

Antigen Thromboxane B2
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Sheep polyclonal antibody raised against Thromboxane B2.
Dilution Radioimmunoassay (1:10000) The optimal working dilution should be determined by the end user.
Host Species Sheep
Immunogen Thromboxane B2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is a sheep polyclonal antibody specific to Thromboxane B2 (TXB2). HAPTEN REACTIVITY: TXB2 100% 6,keto-PGF1alpha 0.17% PGF1alpha 0.14% PGF2alpha 0.13% PGD2 <0.07% 6-KetoPGF1alpha <0.07% 13,14-dihydro-15-keto PGF2alpha <0.07% 13,14-dihydro-6,15diket PGF2alpha <0.07% PGE1 <0.07% PGE3 <0.07% Arachidonic Acid <0.07%
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.