Learn More
Memmert™ Shelf Reversible For 2 Heights (30 Or 60 mm), For Wnb14
Use these shelves with Memmert water baths. Memmert™ Reversible Shelves for Water Baths are available in a wide range of sizes to fit your needs.
Brand:  Memmert™ W14E02895
Additional Details : Weight : 0.01100kg
Product Code. 16374637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Reversible Shelves
  • For Memmert Water Baths
Specifications
Specifications
14 L, 22 L Bath
Reversible Shelf