Learn More
Justrite Shelves for EN Cabinet 226-EN 2903
Click to view available options
Model:
JCBS120S00
JCBS60S000
JCBSROLL00
Length (Metric):
49 cm
109 cm
Unit Size:
Each
Description
- Choice of stainless steel or epoxy coated shelves
- Choice of installed or pull out shelves
Specifications
Specifications
| For Use With (Equipment) | EN Cabinet 226-EN |
| Length (Metric) | 109 cm |
| Model | JCBS120S00 |
| Width (Metric) | 42 cm |
| Color | Light Gray |
| Material | Steel |
| Product Type | Shelf |