Learn More
Justrite Shelves for EN Cabinet 226-EN 2903, Perforated
Additional shelf accessory for EN Cabinets 226-EN. Choice of installed or pull out shelves in stainless steel or epoxy coated finishings.
Brand: Justrite JCBS120LS0
Additional Details : Weight : 1.00000kg
Description
- Choice of stainless steel or epoxy coated shelves
- Choice of installed or pull out shelves
Specifications
Light Gray | |
Shelf | |
Steel | |
109 cm |
Installed | |
42 cm | |
EN Cabinet 226-EN |