missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Shiga toxin 1B Monoclonal Antibody (H82A)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA17099

Product Code. 11504543

  • € 534.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA1-7099 detects the Shiga toxin. This antibody is specific for Shiga toxin type I - B subunit (Stx-1b). MA1-7099 has been successfully used in ELISA procedures. The MA1-7099 immunogen is purified native toxoid.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Shiga toxin 1B
Monoclonal
0.1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA
H82A
Unconjugated
Shiga toxin; Shigatoxin Type 1; Shigatoxin Type I; STX1; STX-1b; STX-Ib
Purified native toxoid.
1 mL
Primary
4°C or -20°C if preferred
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.