missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Shiga toxin 2 subunit B Antibody (028) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26887-100ul
This item is not returnable.
View return policy
Description
Shiga toxin 2 subunit B Monoclonal antibody specifically detects Shiga toxin 2 subunit B in E. coli samples. It is validated for ELISA
Specifications
| Shiga toxin 2 subunit B | |
| Monoclonal | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| E. coli | |
| Purified |
| ELISA | |
| 028 | |
| ELISA 1:5000-1:10000 | |
| Shiga toxin 2 B subunit, Shiga toxin 2B, Shiga toxin 2d subunit B, Stx2 holotoxin B subunit, stx2B, stx2gB, Stx2-NV206 toxin B | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Enterohemorrhagic E. coli (EHEC) Shiga toxin 2 subunit B (Uniprot#: Q93EY4; Met1-Asp89). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction