missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shiga toxin subunit B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 208.00 - € 358.00
Specifications
| Antigen | Shiga toxin subunit B |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655128
|
Novus Biologicals
NBP3-12726-100ul |
100 μg |
€ 358.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640277
|
Novus Biologicals
NBP3-12726-50ul |
50 μg |
€ 208.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Shiga toxin subunit B Polyclonal antibody specifically detects Shiga toxin subunit B in E. coli samples. It is validated for ELISASpecifications
| Shiga toxin subunit B | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| E. coli | |
| Flags: Precursor, Shiga toxin subunit B | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| Produced in rabbits immunized with purified, recombinant E. coli Shiga toxin subunit B (Uniprot#: Q93EY4; Met1-Asp89). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title