missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Shiga toxin subunit B Antibody Pair [HRP]

Product Code. 18634701 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18634701

Brand: Novus Biologicals™ NBP2794215Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for E. coli Shiga toxin subunit B.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 34.38 pg/mL
Assay Sensitivity 34.38-2200 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for E. coli Shiga toxin subunit B.
Target Species E. coli
For Use With (Application) Sandwich ELISA
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.2 mg/mL of mouse anti-Shiga toxin subunit B monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Capture Antibody: 1 mg/mL of mouse anti-Shiga toxin subunit B monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 50 ng of recombinant Shiga toxin subunit B.
  • Quantity 5 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Shiga toxin subunit B
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in dilution buffer before use. 3. Standard: Reconstitute with 1 ml dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2200 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.