missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Shigella Antibody (306/305), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62918H
This item is not returnable.
View return policy
Description
Shigella Monoclonal specifically detects Shigella in Bacteria samples. It is validated for ELISA.
Specifications
| Shigella | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This product is specific for Shigella flexneri serotype 6. S. flexneri is a gram negative, rod shaped bacteria that is very closely related to E. coli and Salmonella. | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA | |
| 306/305 | |
| ELISA | |
| Shigella Boydii; Shigella Dysenteriae; Shigella Flexneri; Shigella Sonnei | |
| NCTC 9729 | |
| 0.1 mL | |
| Primary | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction