missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHIP Antibody (SHIP-01), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-81538UV
This item is not returnable.
View return policy
Description
SHIP Monoclonal antibody specifically detects SHIP in Human samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular), CyTOF
Specifications
| SHIP | |
| Monoclonal | |
| DyLight 350 | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140 | |
| Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Signal Transduction | |
| 3635 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| SHIP-01 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction