missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHISA4 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: RQQLQSPFEG QEIPMTGIPV QPVYPYPQDP QAGPA Highest antigen sequence indentity to the following orthologs: Mouse - 86%, Rat - 83%.
SHISA4 plays an essential role in the maturation of presomitic mesoderm cells by individual attenuation of both FGF and WNT signaling. The Shisa family of single-transmembrane proteins is characterized by an N-terminal cysteine-rich domain and a proline-rich C-terminal region. Its founding member, Xenopus Shisa, promotes head development by antagonizing Wnt and FGF signaling. Shisa physically interacted with immature forms of the Wnt receptor Frizzled and the FGF receptor within the ER and inhibited their posttranslational maturation and trafficking to the cell surface. Loss of Shisa function sensitized the neuroectoderm to Wnt signaling and suppressed head formation during gastrulation.
Specifications
Specifications
| Antigen | SHISA4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SHISA4 |
| Gene Accession No. | Q96DD7 |
| Gene Alias | 9330132O05Rik; C1orf40; mShisa4; Protein shisa-4; shisa family member 4; shisa homolog 4; SHISA4; Tmem58; transmembrane protein 58; UNQ583/PRO1153 |
| Gene Symbols | SHISA4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?