missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SHISA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563631
This item is not returnable.
View return policy
Description
Immunogen sequence: VGAKVAFSKA SRAPRAHRDI NVPRALVDIL RHQA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
The SHISA7 gene encodes a member of the Shisa family proteins, which are involved in the regulation of signaling pathways critical for embryonic development and cellular communication. SHISA7 is particularly influential in modulating synaptic transmission in the central nervous system. It plays a key role in regulating the surface expression and activity of AMPA-type glutamate receptors, which are crucial for fast excitatory neurotransmission and synaptic plasticity in the brain. By controlling the trafficking and localization of these receptors, SHISA7 impacts synaptic strength and neuronal excitability, affecting learning, memory, and other cognitive functions. The functional regulation by SHISA7 of AMPA receptors can have implications for neuropsychiatric disorders, where dysregulation of synaptic transmission is a common feature. Understanding SHISA7's role in synaptic modulation could lead to insights into the mechanisms underlying cognitive processes and the development of therapeutic strategies for neurological conditions.
Specifications
| SHISA7 | |
| Polyclonal | |
| Unconjugated | |
| SHISA7 | |
| CKAMP59; Cystine-knot AMPAR modulating protein of 59 kDa; hCG_2039146; Protein shisa-6-like; Protein shisa-7; shisa family member 7; shisa homolog 7; SHISA7; UPF0626 protein A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 729956 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A6NL88 | |
| SHISA7 | |
| Recombinant protein corresponding to Human SHISA7. Recombinant protein control fragment (Product #RP-101586). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction