missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHISA9 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76496B
This item is not returnable.
View return policy
Description
SHISA9 Polyclonal antibody specifically detects SHISA9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SHISA9 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| CKAMP44, cystine-knot AMPAR modulating protein, DKFZp686D24206, DKFZp686E24206, FLJ10015, FLJ37653, FLJ58277, FLJ99079, protein shisa-9, shisa homolog 9 (Xenopus laevis) | |
| Antibody was raised against a 22 amino acid synthetic peptide near the center of human SHISA9. The immunogen is located within amino acids 120 - 170 of SHISA9. Amino Acid Squence: NTGKPPARKDDPLHDPTKDKT | |
| 0.1 mL | |
| Signal Transduction | |
| 729993 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction